Merck
  • An electrochemical acetylcholine sensor based on lichen-like nickel oxide nanostructure.
An electrochemical acetylcholine sensor based on lichen-like nickel oxide nanostructure.
Biosensors & bioelectronics (2013-05-21)
N Sattarahmady, H Heli, R Dehdari Vais
ABSTRACT
Lichen-like nickel oxide nanostructure was synthesized by a simple method and characterized. The nanostructure was then applied to modify a carbon paste electrode and for the fabrication of a sensor, and the electrocatalytic oxidation of acetylcholine (ACh) on the modified electrode was investigated. The electrocatalytic efficiency of the nickel oxide nanostructure was compared with nickel micro- and nanoparticles, and the lichen-like nickel oxide nanostructure showed the highest efficiency. The mechanism and kinetics of the electrooxidation process were investigated by cyclic voltammetry, steady-state polarization curve and chronoamperometry. The catalytic rate constant and the charge transfer coefficient of ACh electrooxidation by the active nickel species, and the diffusion coefficient of ACh were reported. A sensitive and time-saving hydrodynamic amperometry method was developed for the determination of ACh. ACh was determined with a sensitivity of 392.4 mA M⁻¹ cm⁻² and a limit of detection of 26.7 μM. The sensor had the advantages of simple fabrication method without using any enzyme or reagent and immobilization step, high electrocatalytic activity, very high sensitivity, long-term stability, and antifouling surface property toward ACh and its oxidation product.
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%
Sigma-Aldrich
Nickel(II) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, 99.99% trace metals basis