Merck
  • Direct synthesis of unprotected phenols using palladium-catalysed cross coupling reactions of functionalised organozinc reagents.
Direct synthesis of unprotected phenols using palladium-catalysed cross coupling reactions of functionalised organozinc reagents.
Organic & biomolecular chemistry (2004-01-23)
Richard F W Jackson, Ian Rilatt, P John Murray
ABSTRACT
Palladium-catalysed reaction of unprotected 2-, 3-, and 4-iodophenols with a range of amino acid derived organozinc reagents (not used in excess) gives the expected products in good to excellent yield, demonstrating that carbon-zinc bonds are not protonated by acidic phenols under the conditions of palladium-catalysed coupling reactions.
Sigma-Aldrich
3-Iodobenzyl alcohol, 99%
Pricing and availability is not currently available.