Merck
The type-I interferon receptor. The long and short of it.
Cytokine & growth factor reviews (1996-08-01)
P Domanski, O R Colamonici
ABSTRACT
The type-I interferon receptor is a multisubunit receptor of the cytokine receptor superfamily. The production of specific monoclonal antibodies against the receptor and the cloning of different receptor subunits have contributed to understanding the type-I interferon receptor structure and function. The present article analyzes these new advances and the role of the different receptor subunits in type-I interferon signaling.