Merck
  • Facile synthesis of (Z)-tetracos-5-enoic acid and racemic cis-4-(2-octadecylcyclopropane-1-yl)-butanoic acid.
Facile synthesis of (Z)-tetracos-5-enoic acid and racemic cis-4-(2-octadecylcyclopropane-1-yl)-butanoic acid.
Chemistry and physics of lipids (2003-10-29)
Geoffrey D Coxon, James D Douglas, David E Minnikin
ABSTRACT
(Z)-tetracos-5-enoic acid and racemic cis-4-(2-octadecylcyclopropane-1-yl)-butanoic acid have been prepared from 1-eicosene by a new facile route. Periodic acid cleavage of the epoxide of 1-eicosene gave nonadecanal which was condensed with 4-carboxybutyltriphenylphosphonium bromide to give predominately (Z)-tetracos-5-enoic acid. Simmons-Smith type cyclopropanation of (Z)-tetracos-5-enoic acid gave a minor proportion of racemic cis-4-(2-octadecylcyclopropane-1-yl)-butanoic acid accompanied by major amounts of its methyl ester.