Merck
Epidermal growth factor receptor in tumor angiogenesis.
Hematology/oncology clinics of North America (2004-10-12)
Lee M Ellis
ABSTRACT
This article focuses on the preclinical evidence for activation of the epidermal growth factor receptor (EGFR) in promoting angiogenesis and the efficacy of anti-EGFR agents in inhibiting angiogenesis and tumor growth.
Sigma-Aldrich
VEGF165, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
VEGF121, recombinant, expressed in HEK 293 cells, suitable for cell culture