- Cement luting thickness beneath porcelain veneers made on platinum foil.
Cement luting thickness beneath porcelain veneers made on platinum foil.
The Journal of prosthetic dentistry (1992-09-01)
J G Wall, M H Reisbick, K G Espeleta
PMID1432759
ABSTRACT
Porcelain laminate veneers were made using a platinum foil matrix and were subsequently cemented to mandibular anterior Cymel teeth. Cement film thickness was measured in six predetermined locations. Repeated measures analysis of variance and single degree of freedom contrasts delineated a significant difference between marginal openings at the incisal edge where foil is folded and in four of the other vertical areas (132 versus 74.1 microns). Marginal cement film thickness of veneers made on platinum foil is less than that reported for veneers made on a refractory investment.
Product Description
Platinum, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Platinum, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 0.1m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Platinum, foil, 0.05m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, not light tested, 50x50mm, thickness 0.003mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.003mm, as rolled, 99.95%
Platinum, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.01mm, as rolled, 99.95%
Platinum, foil, 8mm disks, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 0.2m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 6mm disks, thickness 0.0125mm, as rolled, 99.95%
Platinum, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 0.2m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%
Platinum, foil, 4mm disks, thickness 0.05mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 8mm, as rolled, 99.95%
Platinum, foil, not light tested, 25x25mm, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%