Merck
Phenotyping of human complement component C4, a class-III HLA antigen.
The Biochemical journal (1986-11-01)
E Sim, S J Cross
ABSTRACT
The plasma complement protein C4 is encoded at two highly polymorphic loci, A and B, within the class-III region of the major histocompatibility complex. At least 34 different polymorphic variants of human C4 have been identified, including non-expressed or 'null' alleles. The main method of identification of C4 polymorphic allotypes is separation on the basis of charge by agarose-gel electrophoresis of plasma. On staining by immunofixation with anti-C4 antibodies, each C4 type gives three major bands, but, since individuals can have up to five allotypes, the overlapping banding pattern is difficult to interpret. We show that digestion of plasma samples with carboxypeptidase B, which removes C-terminal basic amino acids, before electrophoresis, produces a single, sharp, distinct band for each allotype and allows identification of the biochemical basis of the multiple banding pattern previously observed in C4 phenotype determination.