Merck
  • Determination of felodipine, its enantiomers, and a pyridine metabolite in human plasma by capillary gas chromatography with mass spectrometric detection.
Determination of felodipine, its enantiomers, and a pyridine metabolite in human plasma by capillary gas chromatography with mass spectrometric detection.
Journal of chromatography. B, Biomedical applications (1995-04-21)
J D Dru, J Y Hsieh, B K Matuszewski, M R Dobrinska
ABSTRACT
Sensitive methods based on capillary gas chromatography (GC) with mass spectrometric (MS) detection in a selected-ion monitoring mode (SIM) for the determination of racemic felodipine, its enantiomers, and a pyridine metabolite in human plasma are described. Following liquid-liquid extraction from plasma, enantiomers of felodipine were separated on a chiral HPLC column (Chiralcel OJ) and fractions containing each isomer were collected on a continuous basis using a fraction collector. These fractions were later analyzed by GC-MS-SIM. A similar method based on GC-MS-SIM detection was developed for the determination of racemic felodipine and its pyridine metabolite with a minor modification of sample preparation. The limits of quantitation in plasma were 0.1 ng/ml for both the R(+)- and S(-)-enantiomers of felodipine and 0.5 ng/ml for both racemic felodipine and its pyridine metabolite. The stereoselective assay was used to support a clinical study with racemic felodipine, and was capable of analyzing more than 30 plasma samples per day.
Product Description
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)