Merck
  • Markers of dopamine depletion and compensatory response in striatum and cerebrospinal fluid.
Markers of dopamine depletion and compensatory response in striatum and cerebrospinal fluid.
Journal of neural transmission. Parkinson's disease and dementia section (1995-01-01)
D A Loeffler, P A LeWitt, A J DeMaggio, P L Juneau, P E Milbury, W R Matson
ABSTRACT
Though depletion of CSF homovanillic acid (HVA) concentration has often been regarded as a direct indicator of dopamine (DA) deficiency in Parkinson's Disease (PD), CSF HVA is normal in mildly affected patients. To explore why, we measured DA and its metabolites in striatum and CSF in rabbits receiving reserpine for 5 days. Reserpine, which depletes striatal DA by disrupting vesicular storage of the neurotransmitter, results in a compensatory increase of DA turnover. In response to a 96% depletion of striatal DA, its catabolic intermediates 3,4-dihydroxyphenylacetic acid (DOPAC) and 3-methoxytyramine (3-MT) decreased 64% and 92% in striatum, although the endproduct, HVA, was unchanged. In contrast, CSF concentrations of HVA and DOPAC increased significantly, though 3-MT and levodopa (LD) were unaltered. A 5-fold rise in striatal LD concentration after reserpine-induced DA depletion provided evidence for enhanced DA synthesis. As in PD, the compensatory increase of DA synthesis after reserpine administration confounds the ability of CSF HVA to reflect DA depletion.
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm wire