Merck
  • A gas chromatographic--mass spectrometric study of profiles of volatile metabolites in hepatic encephalopathy.
A gas chromatographic--mass spectrometric study of profiles of volatile metabolites in hepatic encephalopathy.
Journal of chromatography (1981-12-11)
E M Goldberg, L M Blendis, S Sandler
ABSTRACT
Volatile organic substances present in blood plasma and cerebrospinal fluids of certain control groups of human subjects and cirrhotic patients some of whom were suffering from hepatic encephalopathy were quantitatively analysed and identified. A rapid, reproducible, direct injection capillary column gas chromatographic method was developed for the concentration and detection of such volatiles at mg/l and lower concentrations. Of at least forty volatiles detected, twenty-one were identified. The mean concentration of one of these, 3-methylbutanal, was found to be significantly elevated (p less than 0.01) in chronic encephalopathics (2.37 +/- 0.79 mg/l, n = 18), when compared to the controls (0.30 +/- 0.08 mg/l, n = 20). Furthermore, the concentration of this component increased with the clinically diagnosed severity of the encephalopathic state. The presence of 3-methylbutanal is related to leucine, a branched-chain amino acid linked with hepatic encephalopathy.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
SAFC
L-Valine
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Isovaleraldehyde, 97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Isovaleraldehyde, natural, ≥95%, FG
Sigma-Aldrich
Isovaleraldehyde, ≥97%, FG
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%