Merck
  • Folate and methylation status in relation to phosphorylated tau protein(181P) and beta-amyloid(1-42) in cerebrospinal fluid.
Folate and methylation status in relation to phosphorylated tau protein(181P) and beta-amyloid(1-42) in cerebrospinal fluid.
Clinical chemistry (2007-03-27)
Rima Obeid, Mariz Kasoha, Jean-Pierre Knapp, Panagiotis Kostopoulos, George Becker, Klaus Fassbender, Wolfgang Herrmann
ABSTRACT
Increased plasma total homocysteine (tHcy) is a risk factor for neurological diseases, but the underlying pathophysiology has not been adequately explained. We evaluated concentrations of tHcy, S-adenosyl homocysteine (SAH), S-adenosyl methionine (SAM), folate, and vitamin B(12) in cerebrospinal fluid (CSF) and plasma or serum from 182 patients with different neurological disorders. We measured concentrations of phosphorylated tau protein (P-tau)((181P)) and beta-amyloid(1-42) in the CSF. Aging was associated with higher concentrations of tHcy and SAH in the CSF, in addition to lower concentrations of CSF folate and lower SAM:SAH ratio. Concentrations of CSF SAH and CSF folate correlated significantly with those of P-tau (r = 0.46 and r = -0.28, respectively). Moreover, P-tau correlated negatively with SAM:SAH ratio (r = -0.40, P <0.001). The association between SAH and higher P-tau was observed in 3 age groups (<41, 41-60, and >60 years). CSF tHcy was predicted by concentrations of CSF cystathionine (beta = 0.478), folate (beta = -0.403), albumin (beta = 0.349), and age (beta = 0.298). tHcy concentration in the brain is related to age, B vitamins, and CSF albumin. Increase of CSF SAH is related to increased CSF P-tau; decreased degradation of P-tau might be a plausible explanation. Disturbed methyl group metabolism may be the link between hyperhomocysteinemia and neurodegeneration. Lowering tHcy and SAH might protect the brain by preventing P-tau accumulation.
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Titanium, IRMM®, certified reference material, 0.5 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis