Merck
Zinc homeostasis in aging: two elusive faces of the same "metal".
Rejuvenation research (2006-05-19)
Eugenio Mocchegiani, Laura Costarelli, Robertina Giacconi, Catia Cipriano, Elisa Muti, Lothar Rink, Marco Malavolta
ABSTRACT
Proteins involved in zinc homeostasis may be altered in aging. This phenomenon may lead to zinc deficiency in the peripheral blood and an accumulation of zinc bound to insoluble aggregates at the extracellular level in the brain. Therefore, it should be more correct to talk about aging as a condition associated with zinc dyshomeostasis rather than deficiency. Restoring functional zinc homeostasis in aging people is an attractive field for antiaging research, but requires further knowledge than the current state of the art.
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, mossy, ≥99%