Merck
  • International Conference on Harmonisation; final recommendations on the revision of the permitted daily exposures for two solvents, n-methylpyrrolidone and tetrahydrofuran, according to the maintenance procedures for the guidance Q3C Impurities: Residual Solvents; Availability. Notice.
International Conference on Harmonisation; final recommendations on the revision of the permitted daily exposures for two solvents, n-methylpyrrolidone and tetrahydrofuran, according to the maintenance procedures for the guidance Q3C Impurities: Residual Solvents; Availability. Notice.
Federal register (2003-11-19)
ABSTRACT
The Food and Drug Administration (FDA) is announcing final recommendations to revise the permitted daily exposures (PDEs) for two solvents, n-methylpyrrolidone (NMP) and tetrahydrofuran (THF), according to the maintenance procedures for the guidance for industry entitled "Q3C Impurities: Residual Solvents." The final recommendations were reached under the auspices of the International Conference on Harmonisation of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH).
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%