Merck
  • L-lactic acid as a mosquito (Diptera: Culicidae) repellent on human and mouse skin.
L-lactic acid as a mosquito (Diptera: Culicidae) repellent on human and mouse skin.
Journal of medical entomology (2001-03-28)
Y Shirai, K Kamimura, T Seki, M Morohashi
ABSTRACT
The attraction of Aedes albopictus (Skuse) to hands and forearms of human subjects treated with several concentrations of L-LA solution were studied in a test chamber containing proboscis-amputated mosquitoes. Fewer mosquitoes alighted on L-LA treated human skin than on water-treated control skin. Similar results were found using normal mosquitoes following L-LA and water treatment of mouse skin. The relative repellent effects of L-LA varied with concentration. The minimum repellent concentration was lower than previously reported for human skin. The number of alightments decreased at increasing concentrations of L-LA, demonstrating the absolute repellency of L-LA. Unlike previous reports suggesting that L-LA attracted mosquitoes, our studies using human and mouse skin showed that L-LA exhibited both relative and absolute repellency.
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sigma-Aldrich
L-(+)-Lactic acid, 80%
Sigma-Aldrich
L-(+)-Lactic acid, BioXtra, ≥98% (titration)
Sigma-Aldrich
L-(+)-Lactic acid, ≥98%