Merck
  • Riboflavin and riboflavin-derived cofactors in adolescent girls with anorexia nervosa.
Riboflavin and riboflavin-derived cofactors in adolescent girls with anorexia nervosa.
The American journal of clinical nutrition (1999-04-10)
C D Capo-chichi, J L Guéant, E Lefebvre, N Bennani, E Lorentz, C Vidailhet, M Vidailhet
ABSTRACT
Thyroid hormones, riboflavin, riboflavin cofactors, and organic acids were assessed in girls with anorexia nervosa. The objective was to examine the effect of malnutrition and low thyroid hormone concentrations on erythrocyte and plasma riboflavin metabolism and their relation with urinary organic acid excretion. Seventeen adolescent girls with anorexia nervosa [body mass index (BMI; in kg/m2): 14.8 +/- 2.2] and 17 age-matched, healthy girls (control subjects; BMI: 20.5 +/- 2.2) took part in the feeding study. Erythrocyte and plasma riboflavin as well as riboflavin cofactors (flavin mononucleotide and flavin adenine dinucleotide) were assessed by HPLC, whereas urinary organic acids were assessed by gas chromatography-mass spectrometry. Anorectic patients who began a feeding program had higher erythrocyte riboflavin (3.5 +/- 2.2 compared with <0.1 nmol/mol hemoglobin; P < 0.001), lower plasma flavin adenine dinucleotide (57.8 +/- 18.5 compared with 78.5 +/- 54.3 nmol/L; P < 0.05), and higher urinary ethylmalonic acid (7.12 +/- 4.39 compared with 1.3 +/- 2.8 micromol/mmol creatinine; P < 0.001) and isovalerylglycine (7.65 +/- 4.78 compared with 3.8 +/- 0.9 micromol/mmol creatinine; P < 0.05) concentrations than did control subjects. Triiodothyronine concentrations were low and negatively correlated with plasma riboflavin concentrations (r = -0.69, P < 0.01). Not all patients showed improvements in these biochemical indexes after 30 d of refeeding. The low triiodothyronine concentrations observed in anorexia nervosa could alter the extent of riboflavin conversion into cofactors, thus leading to high erythrocyte riboflavin concentrations, low plasma flavin adenine dinucleotide concentrations, and high rates of ethylmalonic acid and isovalerylglycine excretion.
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Supelco
Glutaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glutaric acid, 99%
Sigma-Aldrich
Suberic acid, 98%
Sigma-Aldrich
Ethylmalonic acid, 97%
Sigma-Aldrich
Adipic acid, 99%
Sigma-Aldrich
Suberic acid, purum, ≥98.0% (T)
Supelco
Adipic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Adipic acid, BioXtra, ≥99.5% (HPLC)