Merck
  • Corticotropin-releasing factor triggers neurite outgrowth of a catecholaminergic immortalized neuron via cAMP and MAP kinase signalling pathways.
Corticotropin-releasing factor triggers neurite outgrowth of a catecholaminergic immortalized neuron via cAMP and MAP kinase signalling pathways.
The European journal of neuroscience (2001-04-12)
G Cibelli, P Corsi, G Diana, F Vitiello, G Thiel
ABSTRACT
Corticotropin-releasing factor (CRF), a neuropeptide of 41 amino acids, acts as the major physiological regulator of the basal and stress-induced release of corticotropin (ACTH), beta-endorphin and other proopiomelanocortin-derived peptides from the anterior pituitary gland. In addition to its endocrine activity, CRF displays extrahypophysiotropic effects, mainly as a regulator of stress responses. We show here that CRF may additionally function as a differentiating factor in immortalized noradrenergic neuronal CATH.a cells that express CRF receptor type I and resemble locus coeruleus-derived neurons. CRF triggers morphological changes in CATH.a cells including the appearance of extended long, slender neurites with prominent growth cones. CRF-treated CATH.a cells exhibit a morphology similar to locus coeruleus neurons in primary culture. CRF-induced neurite outgrowth of CATH.a cells was blocked by addition of inhibitors for cAMP-dependent protein kinase or extracellular signal-regulated protein kinase (ERK), a subtype of the mitogen-activated protein kinases. The participation of ERK within the CRF signalling cascade was further confirmed by Western blot experiments, with antibodies directed against the phosphorylated form of ERK, and also with transcription-based assays. We conclude that CRF functions as a differentiating factor of CATH.a cells via the cAMP and the MAP kinase signalling pathways.