Merck
Synthesis of azasugars as potent inhibitors of glycosidases.
Bioorganic & medicinal chemistry (1997-03-01)
Y Le Merrer, L Poitout, J C Depezay, I Dosbaa, S Geoffroy, M J Foglietti
ABSTRACT
A series of enantiomerically pure azasugars (2,5-dideoxy-2, 5-imino-D-mannitol, 1-deoxynojirimycin, 1-deoxymannojirimycin, and related compounds) was synthesized from D-mannitol via aminoheterocyclization of C2-symmetric bis-epoxides and subsequently followed by ring isomerization in few cases. These compounds have been evaluated as inhibitors of several glycosidases (alpha- and beta-D-glucosidases, alpha-D-mannosidase and alpha-L-fucosidase). Inhibition studies indicate notably that the polyhydroxylated azepanes are inhibitors of glycosidases, with Ki in the micromolar range.