Merck
  • Demonstration of initiation potential of carcinogens by induction of preneoplastic glutathione S-transferase P-form-positive liver cell foci: possible in vivo assay system for environmental carcinogens.
Demonstration of initiation potential of carcinogens by induction of preneoplastic glutathione S-transferase P-form-positive liver cell foci: possible in vivo assay system for environmental carcinogens.
Japanese journal of cancer research : Gann (1993-03-01)
H Tsuda, K Matsumoto, H Ogino, M Ito, I Hirono, M Nagao, K Sato, R Cabral, H Bartsch
ABSTRACT
In a development trial for an initiation bioassay system, 7 known carcinogens and 1 suspected carcinogen were examined. In experiment 1, group 1 animals were initially subjected to partial hepatectomy (PH) 12 h before administration of diethylnitrosamine, 2-amino-3-methylimidazo[4,5-f]-quinoline (IQ), captafol, alpha-hexachlorocyclohexane or diethylstilbestrol (DES), then 2 weeks later underwent a promotion procedure comprising administration of phenobarbital (0.05% in diet) for 8 weeks and D-galactosamine (300 mg/kg, i.g.) at week 3. Group 2 received the promotion protocol alone as in group 1. Initiating potential was assayed on the basis of significant increase in values of preneoplastic placental form glutathione S-transferase-positive (GST-P+) foci of more than 3 cells in cross section at week 10. Numbers and areas of GST-P+ foci in group 1 given IQ, captafol and DES were significantly increased as compared to group 2, confirming the validity of the protocol as an initiation assay. In Experiment 2, group 1 rats were subjected to PH and 12 h later received a suspected carcinogenic mixture of opium pyrolysate (OP) or carcinogenic pesticide p,p'-dichloro-diphenyltrichloroethane or hexachlorobenzene. Application of a modified promotion procedure comprising cholic acid (0.15%) and carbon tetrachloride (1 ml/kg, i.g.) revealed significant initiation potential for OP. Overall the results indicate that the current protocols may be useful for detection of the initiation potential of carcinogens irrespective of their mutagenicity.