Merck
  • The assay and absorption kinetics of oral theophylline-7-acetic acid in the human.
The assay and absorption kinetics of oral theophylline-7-acetic acid in the human.
Biopharmaceutics & drug disposition (1981-04-01)
S Sved, I J McGilveray, N Beaudoin
ABSTRACT
A sensitive and specific method for the estimation of theophylline-7-acetic acid in plasma by high performance liquid chromatography is described. Acidified plasma is extracted with chloroform-n-butanol (85: 15), back extracted with phosphate buffer (0.5 mmol-1, pH 6.5), and finally the acidified aqueous phase extracted again with chloroform-butanol. After evaporation of the extract the residue is redissolved in the chromatographic mobile phase (hexane-dichloromethane-ethanol-acetic acid, 10 : 86 : 3 : 1) and chromatographed on silica gel. The method was used to follow the plasma theophylline-7-acetic acid concentrations in two volunteers after single oral doses of 1.0 g of the drug. The results confirm previous reports, based on urine data and on plasma data following intravenous administration, that the drug is poorly absorbed (apparent bioavailability of 1 and 2 per cent), rapidly eliminated (half-life of 1.0 and 2.4 h) and not converted to theophylline.