- New fluorochromes for thiols: maleimide and iodoacetamide derivatives of a 3-phenylcoumarin fluorophore.
New fluorochromes for thiols: maleimide and iodoacetamide derivatives of a 3-phenylcoumarin fluorophore.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1981-02-01)
T O Sippel
PMID7019305
ABSTRACT
N-(4-(7 - Diethylamino - methylcoumarin - 3 yl)phenyl)maleimide (CPM) and the corresponding iodoacetamide are described. When applied to sections at pH 6 and 9, respectively, the two fluorogens are very similar in their reactivity and selectivity toward thiols with which they form adducts having much the same bright blue fluorescence. However, difference in other properties favor CPM for further study.
Sigma-Aldrich
7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin, BioReagent, suitable for fluorescence, ≥85% (HPLC)