- Analysis of propionylpromazine and its metabolites in horse urine.
Analysis of propionylpromazine and its metabolites in horse urine.
The Cornell veterinarian (1984-01-01)
E A Dewey, G A Maylin
PMID6705538
ABSTRACT
The metabolism of propionylpromazine in the horse was studied. Although propionylpromazine is not currently approved or recommended for use in horses, it has been used illegally to alter their performance. Propionylpromazine hydrochloride was administered intramuscularly at clinical and subclinical doses. Three metabolites were detected in urine. The major metabolite was identified as 2-(1-hydroxypropyl) promazine sulfoxide. The detection of this metabolite in routine drug testing has been described.