Merck
[Oxidative degradation of dibenzylsulfide].
Zeitschrift fur allgemeine Mikrobiologie (1979-01-01)
H D Babenzien, I Genz, M Köhler
PMID44589
ABSTRACT
Dibenzylsulfid (DBS) as a model of the organic sulfur compounds in crude oil was converted by a mixed culture (containing Pseudomonas aeruginosa) into several water soluble organic substances. Whereas these compounds are detectable with DC- and IR-spectroscopic techniques, benzylmercaptoacetic acid (BMA) was the only isolated product of DBS utilization. Efficiency of degradation, respectively, accumulation of BMA were dependent on aeration and pH-regulation.
Sigma-Aldrich
Dibenzyl sulfide, ≥95.0% (HPLC)