- Renal vasodilatation after inhibition of renin or converting enzyme in marmoset.
Renal vasodilatation after inhibition of renin or converting enzyme in marmoset.
The relative importance of angiotensin II for the renal vasodilatory response after converting-enzyme inhibition was evaluated by a comparison of the effects of converting-enzyme and renin inhibition on renal vascular resistance. Renal, mesenteric, and hindquarter blood flows were measured with chronically implanted ultrasonic-pulsed Doppler flow probes in conscious, mildly volume-depleted marmosets after administration of a converting-enzyme inhibitor (enalaprilat, 2 mg/kg iv), a synthetic renin inhibitor (CGP 29,287, 1 mg/kg iv), or a renin-inhibitory monoclonal antibody (R-3-36-16, 0.1 mg/kg iv). Enalaprilat reduced blood pressure (-16 +/- 4 mmHg, n = 6) and induced a selective increase in renal blood flow (27 +/- 8%, n = 6). CGP 29,287 and R-3-36-16 induced comparable reductions in blood pressure (-16 +/- 4 mmHg, n = 6 and -20 +/- 4 mmHg, n = 5, respectively) and selective increases in renal blood flow (36 +/- 12%, n = 6 and 34 +/- 16%, n = 4, respectively). The decrease in renal vascular resistance was of similar magnitude for all of the inhibitors (enalaprilat -28 +/- 3%, CGP 29,287 -32 +/- 6%; and R-3-36-16 -33 +/- 7%). These results indicate that the renal vasodilatation induced after converting-enzyme or renin inhibition is mainly due to decreased formation of angiotensin II.