- Effect of fipexide on passive avoidance behaviour in rats.
Effect of fipexide on passive avoidance behaviour in rats.
Pharmacological research (1989-09-01)
G Serra, M Collu, M C Martellotta, A Forgione, W Fratta
PMID2594616
ABSTRACT
The effect of fipexide, administered at different intervals after the learning trial of a single step-through type passive avoidance situation was studied. The administration of fipexide immediately after the learning trial resulted in a long-lasting facilitation of passive avoidance behaviour. On the contrary, the administration of this compound 1 h prior to the retention test failed to influence passive avoidance behaviour. The results suggest that fipexide facilitates memory consolidation but does not influence retrieval processes.