Merck
  • A randomised controlled trial of the use of aromatherapy and hand massage to reduce disruptive behaviour in people with dementia.
A randomised controlled trial of the use of aromatherapy and hand massage to reduce disruptive behaviour in people with dementia.
BMC complementary and alternative medicine (2013-07-11)
Chieh-Yu Fu, Wendy Moyle, Marie Cooke
ABSTRACT
Aromatherapy and hand massage therapies have been reported to have some benefit for people with dementia who display behavioural symptoms; however there are a number of limitations of reported studies. The aim is to investigate the effect of aromatherapy (3% lavender oil spray) with and without hand massage on disruptive behaviour in people with dementia living in long-term care. In a single blinded randomised controlled trial 67 people with a diagnosis of dementia and a history of disruptive behaviour, from three long-term care facilities were recruited and randomised using a random number table into three groups: (1) Combination (aromatherapy and hand massage) (n = 22), (2) Aromatherapy (n = 23), (3) Placebo control (water spray) (n = 22). The intervention was given twice daily for six weeks. Data on residents' behaviour (CMAI) and cognition (MMSE) were collected before, during and after the intervention. Despite a downward trend in behaviours displayed not one of the interventions significantly reduced disruptive behaviour. Further large-scale placebo controlled studies are required where antipsychotic medication is controlled and a comparison of the methods of application of aromatherapy are investigated. ACTRN12612000917831.
Sigma-Aldrich
Lavender oil 40/42% fleurs, natural, FG
Sigma-Aldrich
Lavender oil, natural, from Lavandula angustifolia L.