Merck
  • 7-Oxo-cholesterol potentiates pro-inflammatory signaling in human M1 and M2 macrophages.
7-Oxo-cholesterol potentiates pro-inflammatory signaling in human M1 and M2 macrophages.
Biochemical pharmacology (2013-04-25)
Brigitta Buttari, Luca Segoni, Elisabetta Profumo, Daniela D'Arcangelo, Stefania Rossi, Francesco Facchiano, Rita Businaro, Luigi Iuliano, Rachele Riganò
ABSTRACT
Macrophages, the major cellular components of atherosclerotic plaques, consist of two main subsets: the pro-inflammatory, M1 or classically activated macrophages, and the anti-inflammatory, M2 or alternatively activated macrophages. The molecular and cellular mechanisms that orchestrate the macrophage polarization and activation that may play a role in plaque progression and stability are poorly understood. Recent studies suggest that oxysterols, oxidative stress-mediated cholesterol oxidation products that are abundant in atherosclerotic lesions, may affect macrophage biology. We investigated whether 7-oxo-cholesterol (7oxo-C) affected polarized human M1 and M2 macrophage phenotypes and functions. Monocyte-derived M1 and M2 macrophages were challenged with 7oxo-C and their phenotype analyzed using flow cytometric analysis, and their function via secretome profiling, the presence of endocytosis and matrix metalloproteinase-9 (MMP-9) release. 7oxo-C increased the expression of HLA-DR in M1 macrophages, and CD14 on M2 macrophages. The oxysterol also reduced CD16 expression on M1 macrophages, while reducing their endocytotic capability and increasing MMP-9 secretion in M2 macrophages. Secretome profiling from cultured cell supernatants showed that 7oxo-C stimulated the production of key pro-atherogenic mediators involved in pro-inflammatory, pro-invasive and pro-angiogenic mechanisms both in M1 and M2 cells. Hypoxic conditions potentiated the effects of 7oxo-C on M1 and M2 cells. The ability of 7oxo-C to polarize macrophages toward a pro-inflammatory state represents a potentially novel mechanism by which oxidative stress can contribute to atherosclerotic lesion progression.
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%