Merck
  • Contribution of moxifloxacin or levofloxacin in second-line regimens with or without continuation of pyrazinamide in murine tuberculosis.
Contribution of moxifloxacin or levofloxacin in second-line regimens with or without continuation of pyrazinamide in murine tuberculosis.
American journal of respiratory and critical care medicine (2013-04-19)
Zahoor Ahmad, Sandeep Tyagi, Austin Minkowski, Charles A Peloquin, Jacques H Grosset, Eric L Nuermberger
ABSTRACT
High-dose levofloxacin (L) (1,000 mg) was as active as moxifloxacin (M) (400 mg) in an early bactericidal activity trial, suggesting these fluoroquinolones could be used interchangeably. Whether pyrazinamide (Z) contributes sterilizing activity beyond the first 2 months in fluoroquinolone-containing second-line regimens remains unknown. We compared the efficacy of M and high-dose L alone or in combination with ethionamide (Et), amikacin (A), and Z given for 2 or 7 months. A pharmacokinetic study was performed to determine the L dose equivalent to 1,000 mg in humans. Treatment started 2 weeks after aerosol infection with Mycobacterium tuberculosis H37Rv. Mice received M or L alone or in combination with 2 months of EtZA followed by 5 months of Et or EtZ. After 2 months of treatment, lung colony-forming unit (CFU) counts were similar in mice receiving either fluoroquinolone alone, but, after 4 and 5 months, CFU counts were 2 log10 lower in mice receiving M. Mice receiving 2MEtZA/3MEt and 2LEtZA/3LEt had 1.0 and 2.7 log10 lung CFUs, respectively. When Z was given throughout, both regimens rendered mice culture negative by 5 months, and most mice did not relapse after 7 months of treatment, with fewer relapses observed in the M group after 6 and 7 months of treatment. In murine tuberculosis, M had superior efficacy compared with L despite lower serum drug exposures and may remain the fluoroquinolone of choice for second-line regimens. Z contributed substantial sterilizing activity beyond 2 months in fluoroquinolone-containing second-line regimens, largely compensating for L's weaker activity.
Product Description
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Levofloxacin, analytical standard