Merck
  • Effects of recombinant human erythropoietin on resistance artery endothelial function in stage 4 chronic kidney disease.
Effects of recombinant human erythropoietin on resistance artery endothelial function in stage 4 chronic kidney disease.
Journal of the American Heart Association (2013-04-16)
Marie Briet, Tlili Barhoumi, Muhammad Oneeb Rehman Mian, Cristina Sierra, Pierre Boutouyrie, Michael Davidman, David Bercovitch, Sharon J Nessim, Gershon Frisch, Pierre Paradis, Mark L Lipman, Ernesto L Schiffrin
ABSTRACT
Recent studies have raised concern about the safety of erythropoiesis-stimulating agents because of evidence of increased risk of hypertension and cardiovascular morbidity and mortality in chronic kidney disease (CKD) patients. In the present study, we investigated the effects of recombinant human erythropoietin (EPO) on endothelial function of gluteal subcutaneous resistance arteries isolated from 17 stage 4 patients (estimated glomerular filtration rate 21.9±7.4 mL/min per 1.73 m(2)) aged 63±13 years. Arteries were mounted on a pressurized myograph. EPO impaired endothelium-dependent relaxation in a concentration-dependent manner. The maximal response to acetylcholine with EPO at 1, 10, and 20 IU/mL was reduced by 12%, 34%, and 43%, respectively, compared with the absence of EPO (P<0.001). EPO-induced endothelial dysfunction was significantly associated with carotid stiffness and history of cardiovascular events. EPO had no effect on norepinephrine-induced vasoconstriction or sodium nitroprusside-induced relaxation. ABT-627, an endothelin type A receptor antagonist, and tempol, a superoxide dismutase mimetic, partially reversed the altered endothelial function in the presence of EPO (P<0.01). Increased expression of endothelin-1 was found in the vessel wall after incubation with EPO. EPO alters endothelial function of resistance arteries in CKD patients via a mechanism involving in part oxidative stress and signaling through an endothelin type A receptor. EPO-induced endothelial dysfunction could contribute to deleterious effects of EPO described in large interventional trials.
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Acetylcholine bromide, ≥99%