Merck
  • High coffee intake, but not caffeine, is associated with reduced estrogen receptor negative and postmenopausal breast cancer risk with no effect modification by CYP1A2 genotype.
High coffee intake, but not caffeine, is associated with reduced estrogen receptor negative and postmenopausal breast cancer risk with no effect modification by CYP1A2 genotype.
Nutrition and cancer (2013-03-28)
Elizabeth C Lowcock, Michelle Cotterchio, Laura N Anderson, Beatrice A Boucher, Ahmed El-Sohemy
ABSTRACT
Associations between caffeine and coffee consumption and breast cancer risk are uncertain, with studies suggesting inverse and null associations. Variation in cytochrome P450 1A2 (CYP1A2), a gene responsible for caffeine metabolism, may modify these associations. Cases (n = 3,062) were recruited through the Ontario Cancer Registry and controls (n = 3,427) through random digit dialing. Logistic regression was used to evaluate associations between breast cancer risk and intakes of 7 caffeine-containing items and total caffeine, and examine whether a genetic variant in CYP1A2 (rs762551) modified these associations. Analyses were stratified by estrogen receptor (ER), menopausal, and smoking status. Generally, coffee and caffeine were not associated with breast cancer risk; however, a significant reduction in risk was observed with the highest category of coffee consumption [≥5 cups per day vs. never, multivariate-adjusted odds ratio (MVOR) = 0.71, 95% confidence interval (CI): 0.51, 0.98]. Variant rs762551 did not modify associations. In stratified analyses, high coffee intake was associated with reduced risk of ER- (MVOR = 0.41, 95% CI: 0.19, 0.92) and postmenopausal breast cancer (MVOR = 0.63, 95% CI: 0.43, 0.94). High coffee consumption, but not total caffeine, may be associated with reduced risk of ER- and postmenopausal breast cancers, independent of CYP1A2 genotype. Further studies are needed to replicate these findings.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard