Merck
  • Dissolution of unmodified waxy starch in ionic liquid and solution rheological properties.
Dissolution of unmodified waxy starch in ionic liquid and solution rheological properties.
Carbohydrate polymers (2013-03-08)
Weiqing Liu, Tatiana Budtova
ABSTRACT
Dissolution of waxy corn starch in 1-ethyl-3-methylimidazolium acetate (EMIMAc) was qualitatively studied and compared with gelatinisation process in water. The rheological properties of starch-EMIMAc solutions were investigated in dilute and semi-dilute regions, from 0.1 to 10 wt% over temperature range from 20 °C to 100 °C. The values of zero shear viscosity were obtained by applying Carreau-Yasuda model to shear-thinning flow curves and plotted vs. polymer concentration. Power law exponents in viscosity-concentration dependence in semi-dilute region were compared with the ones reported previously for microcrystalline cellulose. Intrinsic viscosity was obtained as a function of temperature and compared with the one of microcrystalline cellulose; starch was found to be much less temperature sensitive than cellulose. Amylopectin overlap concentration in EMIMAc was compared with the one in water and 0.5 M NaOH-water. Based on these comparisons it was suggested that starch conformation in EMIMAc is similar to the one in water (compact ellipsoid). The activation energy was calculated for starch-EMIMAc solutions and demonstrated to obey power-law concentration dependence.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh