Merck
  • Overcoming the response plateau in multiple myeloma: a novel bortezomib-based strategy for secondary induction and high-yield CD34+ stem cell mobilization.
Overcoming the response plateau in multiple myeloma: a novel bortezomib-based strategy for secondary induction and high-yield CD34+ stem cell mobilization.
Clinical cancer research : an official journal of the American Association for Cancer Research (2013-01-30)
Ruben Niesvizky, Tomer M Mark, Maureen Ward, David S Jayabalan, Roger N Pearse, Megan Manco, Jessica Stern, Paul J Christos, Lena Mathews, Tsiporah B Shore, Faiza Zafar, Karen Pekle, Zhaoying Xiang, Scott Ely, Donna Skerret, Selina Chen-Kiang, Morton Coleman, Maureen E Lane
ABSTRACT
This phase II study evaluated bortezomib-based secondary induction and stem cell mobilization in 38 transplant-eligible patients with myeloma who had an incomplete and stalled response to, or had relapsed after, previous immunomodulatory drug-based induction. Patients received up to six 21-day cycles of bortezomib plus dexamethasone, with added liposomal doxorubicin for patients not achieving partial response or better by cycle 2 or very good partial response or better (≥VGPR) by cycle 4 (DoVeD), followed by bortezomib, high-dose cyclophosphamide, and filgrastim mobilization. Gene expression/signaling pathway analyses were conducted in purified CD34+ cells after bortezomib-based mobilization and compared against patients who received only filgrastim ± cyclophosphamide. Plasma samples were similarly analyzed for quantification of associated protein markers. The response rate to DoVeD relative to the pre-DoVeD baseline was 61%, including 39% ≥ VGPR. Deeper responses were achieved in 10 of 27 patients who received bortezomib-based mobilization; postmobilization response rate was 96%, including 48% ≥ VGPR, relative to the pre-DoVeD baseline. Median CD34+ cell yield was 23.2 × 10(6) cells/kg (median of 1 apheresis session). After a median follow-up of 46.6 months, median progression-free survival was 47.1 months from DoVeD initiation; 5-year overall survival rate was 76.4%. Grade ≥ 3 adverse events included thrombocytopenia (13%), hand-foot syndrome (11%), peripheral neuropathy (8%), and neutropenia (5%). Bortezomib-based mobilization was associated with modulated expression of genes involved in stem cell migration. Bortezomib-based secondary induction and mobilization could represent an alternative strategy for elimination of tumor burden in immunomodulatory drug-resistant patients that does not impact stem cell yield.