Merck
  • Interruption of hepatocyte growth factor signaling augmented oridonin-induced death in human non-small cell lung cancer A549 cells via c-met-nuclear factor-κB-cyclooxygenase-2 and c-Met-Bcl-2-caspase-3 pathways.
Interruption of hepatocyte growth factor signaling augmented oridonin-induced death in human non-small cell lung cancer A549 cells via c-met-nuclear factor-κB-cyclooxygenase-2 and c-Met-Bcl-2-caspase-3 pathways.
Biological & pharmaceutical bulletin (2012-07-14)
Ying Liu, Qi-Feng Shi, Min Qi, Shin-Ichi Tashiro, Satoshi Onodera, Takashi Ikejima
ABSTRACT
The aim of this study was to elucidate the molecular mechanisms mediating hepatocyte growth factor (HGF)-induced protection against oridonin-induced apoptosis in A549 cells. Oridonin induced decrease in Bcl-2/Bax ratio and activation of caspase-3, while these processes were reversed by HGF, suggesting that HGF played an anti-apoptotic role in oridonin-induced A549 cell death. HGF-induced protective effect was partially attributed to the activation of nuclear factor (NF)-κB and cyclooxygenase 2 (COX-2), since the protective effect was abolished by inhibition of NF-κB or interruption of COX-2. Then the activated COX-2 could prevent cells from initiating the apoptotic response by promoting prostaglandin E₂ (PGE₂) release. Activation of NF-κB-COX-2 by HGF-treatment triggered the increase in Bcl-2/Bax ratio, inhibition of procaspase-3 cleavage, promotion of Ca²⁺-independent intracellular phospholipase A2 (iPLA2) expression and augmentation of PGE₂ release, leading to antagonizing oridonin-induced cell death in A549 cells. HGF-induced cell survival in response to oridonin administration was associated with the activation of c-Met-NF-κB-COX-2 and c-Met-Bcl-2-caspase-3 signaling pathways. iPLA2, downstream effector of caspase-3, also participated in these processes.
Sigma-Aldrich
Oridonin, ≥98% (HPLC), solid