Merck
  • Neuroprotective effects of vanillyl alcohol in Gastrodia elata Blume through suppression of oxidative stress and anti-apoptotic activity in toxin-induced dopaminergic MN9D cells.
Neuroprotective effects of vanillyl alcohol in Gastrodia elata Blume through suppression of oxidative stress and anti-apoptotic activity in toxin-induced dopaminergic MN9D cells.
Molecules (Basel, Switzerland) (2011-06-28)
In Su Kim, Dong-Kug Choi, Hun Jong Jung
ABSTRACT
Gastrodia elata Blume (GE) has long been used in oriental countries as a traditional herbal medicine to relieve symptoms associated with neurological ailments such as vertigo, general paralysis and epilepsy. In this study, we have investigated the effects of GE extracts and its major bioactive components on 1-methyl-4-phenylpyridinium (MPP+)-treated MN9D dopaminergic cells, a classic in vitro model for Parkinson's disease (PD). We found that vanillyl alcohol effectively inhibited the cytotoxicity and improved cell viability in MPP+-induced MN9D dopaminergic cells. The underlying mechanisms of vanillyl alcohol action were also studied. Vanillyl alcohol attenuated the elevation of reactive oxygen species (ROS) levels, decreased in the Bax/Bcl-2 ratio and poly (ADP-ribose) polymerase proteolysis. These results indicate that vanillyl alcohol protected dopaminergic MN9D cells against MPP+-induced apoptosis by relieving oxidative stress and modulating the apoptotic process and is therefore a potential candidate for treatment of neurodegenerative diseases such as Parkinson's disease.
Sigma-Aldrich
Vanillyl alcohol, ≥98%, FG
Sigma-Aldrich
4-Hydroxy-3-methoxybenzyl alcohol, 98%