Merck
  • Design and synthesis of novel insecticides based on the serotonergic ligand 1-[(4-aminophenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]piperazine (PAPP).
Design and synthesis of novel insecticides based on the serotonergic ligand 1-[(4-aminophenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]piperazine (PAPP).
Journal of agricultural and food chemistry (2009-12-17)
Mingyi Cai, Zhong Li, Feng Fan, Qingchun Huang, Xusheng Shao, Gonghua Song
ABSTRACT
1-[(4-Aminophenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]piperazine (PAPP) is a 5-HT(1A) agonist and was reported to display high affinity for serotonin (5-HT) receptor from the parasitic nematode Haemonchus contortus . The present investigation explored the possibility of using PAPP as a lead compound of new insecticides with novel mode of action. On the basis of the PAPP scaffold, a series of 1-arylmethyl-4-[(trifluoromethyl)pyridin-2-yl]piperazine derivatives were designed, synthesized, and evaluated for biological activities against the armyworm Pseudaletia separata (Walker). Bioassays showed that most of the target compounds displayed certain growth-inhibiting activities or larvicidal activities against armyworm. The quantitative structure-activity relationship (QSAR) for growth-inhibiting activities was also analyzed and established.