Merck
Diffusion coefficients of biomolecules using long-lived spin states.
Journal of the American Chemical Society (2009-05-16)
Puneet Ahuja, Riddhiman Sarkar, Paul R Vasos, Geoffrey Bodenhausen
ABSTRACT
We report the first observation of long-lived states (LLS) having lifetimes T(LLS) that exceed the corresponding spin-lattice relaxation times T(1) by more than a factor 6 in a protein. Slow diffusion coefficients characteristic of large biomolecules can be determined by combining LLS methods with moderate pulsed field gradients (PFGs) available on commercial probeheads, as the extension of spin memory reduces the strain on the duration and/or strength of the PFGs. No isotope labeling of the biomolecule is necessary.
Sigma-Aldrich
Ala-Gly
Pricing and availability is not currently available.