Merck
  • Synthesis and anticancer activity of lipophilic platinum(II) complexes of 3,5-diisopropylsalicylate.
Synthesis and anticancer activity of lipophilic platinum(II) complexes of 3,5-diisopropylsalicylate.
European journal of medicinal chemistry (2007-10-26)
Yao Yu, Li-Guang Lou, Wei-Ping Liu, Hua-Jie Zhu, Qing-Song Ye, Xi-Zhu Chen, Wen-Gui Gao, Shu-Qian Hou
ABSTRACT
Novel lipophilic platinum(II) complexes (LSPt-1-3), containing 3,5-diisopropylsalicylate (DIPS) as a leaving group and 2NH(3) or 1R,2R-diaminocyclohexane or (4R,5R)-4,5-bis(aminomethyl)-2-isopropyl-1,3-dioxolane as the carrier, have been synthesized, characterized and evaluated in vitro and in vivo. The octanol/water distribution coefficient of the complexes has also been measured. The results showed that the complexes achieved a typical square planar and the octanol/water distribution coefficient logP was 4.27, 4.37 and 4.31. The complexes were tested by SRB method to be more cytotoxic than Carboplatin, Oxaliplatin and Eptaplatin against 3AO, A549, NCI-H460 and SGC-7901 human cancer cell lines. Among complexes, LSPt-2 was much more effective than Carboplatin and Oxaliplatin in treating the NCI-H460 non-small-cell lung tumor-bearing mice. Its optimal activity was 38.8% (T/C) at a dose of 30 mg/kg following i.p. administration. LD(50) for the complex was found to be 230.9 mg/kg. LSPt-2 exhibited great anticancer activity, good lipophilic ability and low toxicity and therefore, it is a promising candidate for effective and stable pharmaceutical liposomal platinum anticancer drug.
Sigma-Aldrich
3,5-Diisopropyl-2-hydroxybenzoic acid, 98%