Merck
  • 1-Tridecene--male-produced sex pheromone of the tenebrionid beetle Parastizopus transgariepinus.
1-Tridecene--male-produced sex pheromone of the tenebrionid beetle Parastizopus transgariepinus.
Die Naturwissenschaften (2007-09-28)
Sven Geiselhardt, Peter Ockenfels, Klaus Peschke
ABSTRACT
Males of the genus Parastizopus (Coleoptera: Tenebrionidae) exhibit a special pheromone-emitting behaviour. They do a headstand, expose the aedeagus and remain in this posture for a few seconds. The pheromone emitted by P. transgariepinus was collected by solid-phase micro-extraction (100 microm polydimethylsiloxane fibre) and identified as 1-tridecene by gas chromatography/mass spectrometry. Presumably, this compound originates from the aedeagal gland, a special feature in Parastizopus, as 1-tridecene is the main compound in the gland reservoirs (23.6+/-3.8%), accompanied by various less volatile fatty acid esters (25.2+/-2.0%) and hydrocarbons (51.2+/-5.7%). 1-Tridecene is also part of the pygidial defensive secretion of both sexes, together with other 1-alkenes, monoterpene hydrocarbons and 1,4-benzoquinones, but as none of these other compounds was detected during calling, the pygidial gland could be ruled out as pheromone source. Extracts of the aedeagal gland reservoirs and the pygidial defensive secretion contained comparable amounts of 1-tridecene, 1.24+/-0.41 and 1.88+/-0.54 microg/male, respectively. Chemo-orientation experiments using a servosphere showed that 1 microg of 1-tridecene was attractive to females but not to males.
Product Description
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder