Merck
  • Inhibition of TGF-beta induced lung fibroblast to myofibroblast conversion by phosphodiesterase inhibiting drugs and activators of soluble guanylyl cyclase.
Inhibition of TGF-beta induced lung fibroblast to myofibroblast conversion by phosphodiesterase inhibiting drugs and activators of soluble guanylyl cyclase.
European journal of pharmacology (2007-07-31)
Torsten R Dunkern, Daniel Feurstein, Giovanni A Rossi, Federica Sabatini, Armin Hatzelmann
ABSTRACT
Pulmonary fibroblast to myofibroblast conversion is a pathophysiological feature of idiopathic pulmonary fibrosis and COPD. This conversion is induced by transforming growth factor (TGF)-beta derived from epithelial cells as well as activated macrophages that have infiltrated the lung. Preventing this conversion might be a favourable therapeutic approach. Within this study we examined the activity of different members of the phosphodiesterase (PDE) family in primary human lung fibroblasts and various lung fibroblast cell lines both before and after TGF-beta induced differentiation to myofibroblasts as reflected by the expression of alpha-smooth muscle actin. We showed that the predominant PDE activities in lung fibroblasts are attributed to PDE5, PDE1 and to a smaller extent to PDE4. cyclic GMP (cGMP)-hydrolyzing activity declines by about half after differentiation to myofibroblasts in all pulmonary fibroblasts investigated, which is accompanied by a down-regulation of PDE5 protein. Lung fibroblast to myofibroblast differentiation is blocked by treatment with the PDE4 inhibitor piclamilast alone, depending on the TGF-beta concentration applied, and in combination with prostaglandin E(2) (PGE(2)) in a synergistic manner. Despite the high PDE5 activity the PDE5 inhibitor sildenafil by itself as well as in combination with brain natriuretic peptide or the nitric oxide-donor DETA-NONOate shows no inhibiting effects. However, combining sildenafil with the guanylyl cyclase (GC) activator BAY58-2667 and ODQ (which sensitizes GC for activation by BAY58-2667) suppressed TGF-beta induced differentiation. In summary, our data indicate that drugs interfering with the cyclic AMP (cAMP)-as well as with the NO-cGMP-pathway offer the therapeutic opportunity to prevent the differentiation of pulmonary fibroblasts to myofibroblasts in lung fibrosis.
Sigma-Aldrich
Piclamilast, ≥98% (HPLC)