Merck
  • [Role of kappa-opioid receptors in the regulation of cardiac resistance to arrhythmogenic effects of ischemia and reperfusion].
[Role of kappa-opioid receptors in the regulation of cardiac resistance to arrhythmogenic effects of ischemia and reperfusion].
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova (2007-05-26)
A Iu Lishmanov, T V Lasukova, L N Maslov, A A Platonov
ABSTRACT
It was found that pretreatment of rats with selective agonist of kappa1-opioid receptors (OR) (-)--U--50.488 decreased the incidence of ischemic (10 min) and reperfusion (10 min) ventricular arrhythmias. The selective kappa2-OR agonist GR-89696 had no effect on the incidence of ventricular arrhythmias during a 10-min coronary artery occlusion and following reperfusion in anesthetized rats. The effect of (-)--U-50.488 was abolished by the selective kappa1-OR antagonist of non-binaltorphimine and the non-selective peripheral OR antagonist naloxone methiodide. Perfusion of isolated rat heart with (-)--U-50.488 did not affect arrhythmias during ischemia and reperfusion. The authors suggest that stimulation of kappa1-opioid receptors located outside the central nervous system increases heart resistance against arrhythmogenic action of ischemia/reperfusion, antiarrhythmic action of (-)--U-50.488 being mediated through extracardiac opioid receptors.