Merck
  • Effect of food seasoning spices mixture on biomarkers of oxidative stress in tissues of fructose-fed insulin-resistant rats.
Effect of food seasoning spices mixture on biomarkers of oxidative stress in tissues of fructose-fed insulin-resistant rats.
Journal of medicinal food (2007-05-03)
R Suganthi, S Rajamani, M K Ravichandran, C V Anuradha
ABSTRACT
High fructose feeding in normal rats induces insulin resistance and also facilitates oxidative damage. The present study examines the effects of a spices mixture (SM) on oxidative stress markers and antioxidant potential in tissues of high fructose-fed insulin-resistant rats. Male Wistar rats received a semisynthetic diet containing either 60% fructose or 60% starch. SM administration at three different doses (10, 30, and 50 mg/day per rat) was initiated orally 15 days later and continued for the next 30 days. After the total experimental period of 45 days, peroxidation of lipids and antioxidant status in liver and kidney were quantified. Fructose-treated rats showed increased levels of peroxidation indices such as thiobarbituric acid-reactive substances and lipid hydroperoxides in tissues. The condition was associated with an inadequate antioxidant system. Administration of SM along with fructose diet reduced the levels of peroxidation markers in tissues and improved the antioxidant status. The positive effect of SM on the oxidant-antioxidant balance could be attributed to the active constituents of the different spices present in the mixture.