Merck
  • Inhibition of P-type ATPases by [(dihydroindenyl)oxy]acetic acid (DIOA), a K+ -Cl- cotransporter inhibitor.
Inhibition of P-type ATPases by [(dihydroindenyl)oxy]acetic acid (DIOA), a K+ -Cl- cotransporter inhibitor.
European journal of pharmacology (2007-02-17)
Takuto Fujii, Yuta Ohira, Yasuo Itomi, Yuji Takahashi, Shinji Asano, Magotoshi Morii, Noriaki Takeguchi, Hideki Sakai
ABSTRACT
[(Dihydroindenyl)oxy]acetic acid (DIOA) has been used as a potent inhibitor of K+ -Cl- cotransporter (IC(50)=10 microM). Here we found that DIOA inhibited activities of P-type ATPases such as dog kidney Na+,K+-ATPase (IC(50)=53 microM), hog gastric H+,K+-ATPase (IC(50)=97 microM) and rabbit muscle Ca(2+)-ATPase (IC(50)=127 microM). In the membrane preparation of the LLC-PK1 cells stably expressing rabbit gastric H+,K+-ATPase, DIOA inhibited activities of the endogenous Na+,K+-ATPase (IC(50)=95 microM) and the exogenous H+,K+-ATPase (IC(50)=75 microM). 5-Nitro-2-(3-phenylpropylamino)-benzoic acid (NPPB), a Cl- channel blocker, had no effects on the DIOA-elicited inhibition of the P-type ATPases. These findings suggest that lower concentration of DIOA (< 20-30 microM) should be used for evaluation of the activity of K+ -Cl- cotransporter without affecting the activities of coexisting Na+,K+ -ATPase and/or H+,K+-ATPase in cells.
Sigma-Aldrich
R-(+)-DIOA, ≥98% (HPLC), solid
Pricing and availability is not currently available.