Merck
  • Discriminative-stimulus effects of methamphetamine and morphine in rats are attenuated by cAMP-related compounds.
Discriminative-stimulus effects of methamphetamine and morphine in rats are attenuated by cAMP-related compounds.
Behavioural brain research (2006-07-22)
Yijin Yan, Atsumi Nitta, Tomoko Mizuno, Akira Nakajima, Kiyofumi Yamada, Toshitaka Nabeshima
ABSTRACT
Animal models of drug discrimination have been used to examine the subjective effects of addictive substances. The cAMP system is a crucial downstream signaling pathway implicated in the long-lasting neuroadaptations induced by addictive drugs. We examined effects of rolipram, nefiracetam, and dopamine D2-like receptor antagonists, all of which have been reported to modulate cAMP level in vivo, on the discriminative-stimulus effects of methamphetamine (METH) and morphine in rats. All these compounds inhibited the discriminative-stimulus effects of METH, while only rolipram and nefiracetam attenuated the discriminative-stimulus effects of morphine. In addition, neither nifedipine nor neomycin, two voltage-sensitive calcium channel blockers, was found to modulate the effect of nefiracetam on METH-associated discriminative stimuli, suggesting that the inhibitory effect of nefiracetam may not involve the activation of calcium channels. These findings suggest that the cAMP signaling cascade may play a key role in the discriminative-stimulus effects of METH and morphine and may be a potential target for the development of therapeutics to counter drugs of abuse.