Merck
  • The effects of ginsenoside Re and its metabolite, ginsenoside Rh1, on 12-O-tetradecanoylphorbol 13-acetate- and oxazolone-induced mouse dermatitis models.
The effects of ginsenoside Re and its metabolite, ginsenoside Rh1, on 12-O-tetradecanoylphorbol 13-acetate- and oxazolone-induced mouse dermatitis models.
Planta medica (2006-03-25)
Yong-Wook Shin, Eun-Ah Bae, Sung-Soo Kim, Young-Chul Lee, Boo-Young Lee, Dong-Hyun Kim
ABSTRACT
The effects of the main constituent ginsenoside Re in ginseng and its metabolite, ginsenoside Rh1, were investigated in 12-O-tetradecanoylphorbol 13-acetate (TPA)- and oxazolone-induced mouse ear dermatitis models. Ginsenoside Rh1 potently suppressed the TPA- and oxazolone-induced swellings as well as mRNA expression levels of cyclooxygenase-2, IL-1beta and TNF-alpha, although these were only weakly inhibited by ginsenoside Re.
Product Description
Supelco
Ginsenoside Rh1, analytical standard