Merck
  • Identification of components responsible for the odor of cigar smoker's breath.
Identification of components responsible for the odor of cigar smoker's breath.
Journal of agricultural and food chemistry (2006-01-19)
Russell Bazemore, Charles Harrison, Michael Greenberg
ABSTRACT
Following smoking 1/2 of a cigar, the most odorous cigar tobacco smoke components extracted from the surface of the tongue by nylon-meshed swabs and then extracted from the swab headspace by solid phase microextraction were ethyl pyrrole, 2,3-dimethyl pyrazine, and 2-ethyl pyridine. Similar classes of compounds were identified from the headspace of an aqueous simulated saliva solution treated with cigar smoke. The most odorous compounds were 2,3,5-trimethyl pyridine, 2,5-dimethyl pyrazine, and 2-ethyl-3,5-dimethyl pyridine. Pyridines and pyrazines, the most prominent classes of odorous compounds identified in this experiment, may be generated during cigar pyrrolysis by cleavage of nicotine or by Maillard reaction.
Sigma-Aldrich
2-Ethylpyridine, 97%
Pricing and availability is not currently available.