Merck
  • Influence of N,N'-diacetyl-L-cystine on D-galactosamine/lipopolysaccharide induced immunological liver failure in mice.
Influence of N,N'-diacetyl-L-cystine on D-galactosamine/lipopolysaccharide induced immunological liver failure in mice.
Yao xue xue bao = Acta pharmaceutica Sinica (2005-02-11)
Pei Wang, Xing-chang Ren, Jin Yu, Yi Lin, Xi-ming Wu
ABSTRACT
To study the therapeutic effects of N,N'-diacetyl-L-cystine (DiNAC) on immunological liver failure. Serum ALT, AST and T cell subsets in peripheral blood of the experimental animals during the trial period were analyzed by an automatic serum analyzer and a flow cytometer, respectively. The sectioned liver specimens were examined under a light microscope. And 24 h after the injection of Gal/LPS, the survival rate of rats was calculated. DiNAC (50, 200, 800 mg x kg(-1), i.p.) suppressed the elevation of serum levels of ALT and AST, markedly enhanced proliferation and differentiation of T cell subsets (CD4+, CD8+ and Th1, Th2), and improved all the histopathological features. In mice of fulminant hepatic failure (FHF), the survival time significantly prolonged and the survival rate increased 24 h after i.p. DiNAC. These effects were obviously dose-dependent. DiNAC on mice with FHF has an inhibitory action which is related to immune mechanism.
Product Description
Acetylcysteine impurity C, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.