Merck
  • A study of 2-azido-3,5-dichlorobiphenyl by nano- and picosecond laser flash photolysis and computational methods.
A study of 2-azido-3,5-dichlorobiphenyl by nano- and picosecond laser flash photolysis and computational methods.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2004-12-24)
Nina P Gritsan, Dmitrii A Polshakov, Meng-Lin Tsao, Matthew S Platz
ABSTRACT
The dynamics of singlet 3,5-dichloro-2-biphenylnitrene and the products of its rearrangement were monitored by pico- and nanosecond laser flash photolysis and the results were consistent with the predictions of DFT and ab initio molecular orbital calculations.