Merck
  • [Effect of phenothiazine derivative on adrenal cortex response of sheep to repeated emotional stress].
[Effect of phenothiazine derivative on adrenal cortex response of sheep to repeated emotional stress].
Endokrynologia Polska (1991-01-01)
D Wrońska, J Niezgoda, K Pierzchała, A Sechman, S Bobek, A B Hamid
ABSTRACT
The study was aimed at the evaluation of propiopromazine (Combelen, Bayer), a derivative of phenothiazine, as an agent lowering in sheep the response to stress. The stress of emotional origin was induced in sheep by the isolation from herd lasting 1 hour. The isolation experiments were repeated 6 times on the same group of sheep, first three isolations (1-3) in daily intervals and next three (4-6) in weekly intervals. Propiopromazine was administered before each isolation experiment. The reaction of sheep to the isolation stress was weaker after propiopromazine administration. This was suggested by smaller increase in blood serum cortisol and glucose levels when compared to sheep subjected to isolation but not receiving the drug. Such effect was especially conspicuous during the course of the first isolation experiment; during the next experiments the difference concerning the reaction to stress between the sheep isolated from the herd receiving and not receiving the drug was gradually diminishing. It was shown in addition that propiopromazine administration to the sheep not subjected to stress caused an increase in cortisol level by 125 per cent and that in glucose level by 35 per cent. These results suggest that propiopromazine administration protects the organism against the effects of emotional stress only partially. Moreover, the effect of its administration gradually weakens with repeating of the stress inducing experiment, and propiopromazine itself may act as a stress inducing factor. It seems therefore that the use of propiopromazine and similar compounds as anti-stress agents may be questionable.