Merck
  • Spectrum of cross-photosensitization in 18 consecutive patients with contact photoallergy to ketoprofen: associated photoallergies to non-benzophenone-containing molecules.
Spectrum of cross-photosensitization in 18 consecutive patients with contact photoallergy to ketoprofen: associated photoallergies to non-benzophenone-containing molecules.
Contact dermatitis (2003-05-21)
E Durbize, M Vigan, E Puzenat, P Girardin, B Adessi, P H Desprez, P H Humbert, R Laurent, F Aubin
ABSTRACT
Contact photoallergy to ketoprofen gels has been widely reported, and cross-sensitivity reactions with other compounds, such as tiaprofenic acid, fenofibrate and benzophenones, are well known. However, positive photopatch tests to other different non-benzophenone-related compounds have recently been observed. We report the results of photopatch testing in patients with contact photoallergy to ketoprofen and discuss the spectrum of cross-sensitization to ketoprofen. 18 consecutive patients with a history of photocontact dermatitis from ketoprofen were investigated. Patch and photopatch tests were performed. As expected, we observed positive photopatch tests to Ketum* gel and ketoprofen 2.5% in petrolatum in all patients (100%). However, it was remarkable to note positive photopatch tests to other unexpected and non-relevant allergens, including fentichlor (67%), tetrachlorosalicylanilide (28%), triclosan (17%), tribromsalan (11%) and bithionol (11%), with no clinical relevance. Interestingly, these agents belong to the family of halogenated salicylanilides and related compounds, which have been forbidden in Europe since the 1970s. Our results raise the question of hyper-photosusceptibility to non-relevant allergens induced by photosensitivity to ketoprofen. The mechanism may involve the high photoreactivity induced by the association of a benzene ring with an oxygen group.
Product Description
Supelco
Bithionol, VETRANAL®, analytical standard
Pricing and availability is not currently available.