Merck
  • Glycine tranporter-1 blockade potentiates NMDA-mediated responses in rat prefrontal cortical neurons in vitro and in vivo.
Glycine tranporter-1 blockade potentiates NMDA-mediated responses in rat prefrontal cortical neurons in vitro and in vivo.
Journal of neurophysiology (2003-02-08)
Long Chen, Mark Muhlhauser, Charles R Yang
ABSTRACT
The N-methyl-D-aspartate (NMDA) receptor (NMDA-R) has pivotal roles in neural development, learning, memory, and synaptic plasticity. Functional impairment of NMDA-R has been implicated in schizophrenia. NMDA-R activation requires glycine to act on the glycine-B (GlyB) site of the NMDA-R as an obligatory co-agonist with glutamate. Extracellular glycine near NMDA-R is regulated effectively by a glial glycine transporter (GlyT1). Using whole-cell voltage-clamp recordings in prefrontal cortex (PFC) slices, we have shown that exogenous GlyB site agonists glycine and D-serine, or a specific GlyT1 inhibitor N[3-(4'-fluorophenyl)-3-(4'-phenylphenoxy)propyl]sarcosine (NFPS) in the presence of exogenous glycine (10 microM), potentiated synaptically evoked NMDA excitatory postsynaptic currents (EPSCs) in vitro. Furthermore, in urethan-anesthetized rats, microiontophoretic NMDA pulses excite single PFC neurons. When these responses were blocked by approximately 50% to approximately 90% on continuous iontophoretic application of the GlyB site, antagonist (+)HA-966, intravenous NFPS (5 mg/kg), or a GlyB site agonist D-serine (50 mg/kg iv) reversed this (+)HA-966 block. NFPS may elevate endogenous glycine levels sufficiently to displace (+)HA-966 from the GlyB sites of the NMDA-R, thus enabling reactivation of the NMDA-Rs by iontophoretic NMDA applications. D-Serine (50-100 mg/kg iv) or NFPS (1-2 mg/kg iv) alone also augmented NMDA-evoked excitatory responses. These data suggest that direct GlyB site stimulation by D-serine, or blockade of GLYT1 to elevate endogenous glycine to act on unsaturated GlyB sites on NMDA-Rs, potentiated NMDA-R-mediated firing responses in rat PFC. Hence, blockade of GlyT1 to elevate glycine near the NMDA-R may activate hypofunctional NMDA-R, which has been implicated to play a critical role in the pathophysiology of schizophrenia.