Merck
  • Spermine increases the active and passive transport across the alveolar epithelium in situ: effect of thiol reagents.
Spermine increases the active and passive transport across the alveolar epithelium in situ: effect of thiol reagents.
Pflugers Archiv : European journal of physiology (2001-02-24)
G Saumon, G Martet
ABSTRACT
An intact alveolar epithelial barrier is thought to be important for alveolar liquid absorption. However, polycations increase alveolar permeability without affecting alveolar liquid absorption (Saumon et al., Am J Physiol 1995: 269:L185-L194). We have reconsidered this issue using polyamines. The polyamine spermine (10(-3) mol/l) produced a large (up to 20-fold), sustained increase in the permeability of the alveolar barrier to mannitol (PAMan) and in alveolar liquid absorption (Jw, twofold) in isolated rat lungs. These increases were inhibited by 5 x 10(-3) mol/l putrescine and 2 x 10(-3) mol/l spermidine. Because spermine is known to affect the phosphoinositide/Ca2+ signalling pathway, we evaluated the effects of thiol reagents known to interfere with this pathway in different ways. Thimerosal, a thiol reagent which sensitizes the inositol 1,4,5-trisphosphate (IP3) receptor, inhibited the spermine-induced increase in PA(Man) and, to a lesser extent, that of Jw. Mersalyl, a thiol reagent which blocks IP3-gated Ca2+ channels, enhanced spermine's effect, whereas N-ethylmaleimide, a non-specific thiol reagent, had no effect. These observations show that large increases in permeability may coexist with increases in Jw. They also suggest that the phosphoinositide/Ca2+ second messenger pathway is involved in modulating the tightness of the alveolar barrier and alveolar liquid absorption.
Product Description
Supelco
Mersalyl acid, analytical standard